IPTG (isopropyl-β-D-thiogalactopyranoside) (not of animal origin) for biotechnology
Catalog # VWRA06190101
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:238.31 g/mol
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273