Order Entry
India
ContactUsLinkComponent
VWR®, TAQ DNA Polymerase
VWR®, TAQ DNA Polymerase
Catalog # VWRA06105100
Supplier:  VWR Collection
CAS Number:  
VWR®, TAQ DNA Polymerase
Catalog # VWRA06105100
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    1 KU
  • Application:
  • Description:
    TAQ DNA Polymerase, 5U/µL
  • Environmentally Preferable:

Specifications

About this item

VWR® Delivered High quality Reagents & biochemicals, combined with innovative products that gives optimum results , save time & simplify protocols

  • VWR® Lifescience Molecular Biology grade Reagents & Biochemicals