VWR®, Bovine Serum Albumin Heat Shock Fraction V
Catalog # VWRA06190114
Supplier: VWR Collection
CAS Number:
Specifications
- Pk:100 g
- Protease-free:Y
- Protein synonyms:BSA
- Protein/peptide name:Bovine Serum Albumin Heat shock fraction V
- pH:7
Specifications
About this item
VWR® Delivered High quality Reagents & biochemicals, combined with innovative products that gives optimum results , save time & simplify protocols
- VWR® Lifescience Molecular Biology grade Reagents & Biochemicals
essentially globulin free