Hink's TNM-FH medium, Corning®
Catalog # MDTC13-100-CV
Supplier: Corning
CAS Number:
Specifications
- Media Format:Liquid
- Pk:6
- Description:Hink's TNM-FH medium with L-glutamine lactalbumin hydrolysate, and yeastolate, without insect hemolymph
- Environmentally Preferable:
Specifications
About this item
Hink's TNM-FH medium is a modified Grace's formulation containing lactalbumin hydrolysate (LAH) and ultrafiltered yeastolate, and was originally developed for the optimal propagation of cabbage looper, Tricoplusi in cells.