Order Entry
India
ContactUsLinkComponent
HAT (Hypoxanthine, Aminopterin, Thymidine), Corning®
HAT (Hypoxanthine, Aminopterin, Thymidine), Corning®
Catalog # MDTC25-046-CI
Supplier:  Corning
CAS Number:  
undefined
HAT (Hypoxanthine, Aminopterin, Thymidine), Corning®
Catalog # MDTC25-046-CI
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    1
  • Source:
  • Species:
  • Description:
    HAT (Hypoxanthine, Aminopterin, Thymidine)
  • Size:
    100 ml
  • Environmentally Preferable:

Specifications

About this item

HAT Medium (hypoxanthine-aminopterin-thymidine medium) is used in mammalian cell culture for post-fusion selection to eliminate certain types of myeloma cells

The combination of aminopterin, a drug that acts as a powerful folate metabolism inhibitor, with hyproxanthine and thymidine, which are intermediates in DNA synthesis, provides a form of artificial selection for cells containing functioning hypoxanthine-guanine phosphoribosyltransferase (HGPRT) and thymidine kinase (TK). These enzymes are involved in DNA synthesis and cell division. HAT medium is often used in the preparation of monoclonal antibodies.