J.T.Baker® BAKER INSTRA-ANALYZED®, Platinum Standard
Catalog # 6463-04
Supplier: Avantor
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:20% HCl
- Pk:150 mL
- Accuracy:±10 mg/L
- Element:Platinum
- Symbol:Pt
Specifications
About this item
Prepared by dissolution of platinum metal of 99,98% spectral purity
- Traceable to NIST