Order Entry
India
ContactUsLinkComponent
Potassium thiocyanate buffered solution, BAKER ANALYZED®, J.T.Baker®
Irritant
Potassium thiocyanate buffered solution, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
Irritant
Potassium thiocyanate buffered solution, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Appearance (clear, colorless solution):
    Passes Test
  • Potassium Thiocyanate (KSCN), g/100 mL:
    55.3 - 63.6
  • Sodium (Na), g/L:
    2.6 - 3.6
  • Potassium (K), mEQ/L:
    5414 - 6616
  • pH (As Is" basis) at 25*C":
    6.8 - 7.3
  • Chloride Identity (Cl):
    Passes Test
  • Phosphate Identity (PO₄):
    Passes Test
  • Sterility:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together