Order Entry
India
ContactUsLinkComponent
VWR®, Lab Centrifuge
  VWRA521-2823
undefined
VWR®, Lab Centrifuge
  VWRA521-2823
  • Max. speed:
    4500 min-¹ without rotor
  • Temperature:
    Refrigeration –10 °C
  • Description:
    Multipurpose laboratory refrigerated centrifuge
  • Pk:
    1
It is a device which offers powerful, efficient performance with years of durability. The sheer versatility of the device due to a wide range of rotor options make it extemely useful across a range of requiremnets. Moreover, the device can also be programmed easily to match the unique needs of the user with its user-friendly interface. Also the brushless DC motor present in the device is dust-free and guarantees efficient performance without the need for constant maintenance.
  • Microprocessor controlled for efficient performance even under high load
  • Intuitive interface allows for easy and quick setting of parameters
  • Lid lock safety feature to avoid opening of lid when the rotor is running 
  • One touch toggle between RCF and RPM with just a touch of a button
  • Short spin feature for ultra-fast centrifugation and pelleting
  • Adjustable ramp and brake speed
 : Rotors have to be ordered separately.
Rotors