Order Entry
India
ContactUsLinkComponent
VWR®, Microplate Centrifuge with Swing Out Rotor
VWR®, Microplate Centrifuge with Swing Out Rotor
Catalog # VWRA521-2811
Supplier:  VWR Collection
undefined
VWR®, Microplate Centrifuge with Swing Out Rotor
Catalog # VWRA521-2811
Supplier:  VWR Collection

Specifications

  • Max. capacity:
    2× 96/384 microplates
  • Max. speed:
    2200 min⁻¹
  • Description:
    Microplate swing out centrifuge with 2× microplate swing out rotor
  • Max. RCF:
    560 ×g
  • Display:
    Digital
  • Timer:
    1 - 999 mins and infinity mode
  • Weight:
    7,7 kg
  • W×D×H:
    415×355×173 mm
  • Pk:
    1

Specifications

About this item

This microplate centrifuge can accomodate a wide range of PCR and deepwell plates which makes it extremely versatile. Because of the limited space in laboratories, it is designed with a compact footprint to occupy less space. It also has an imbalance detection feature with auto-cut off safety, to ensure smooth functioning.

  • Microprocessor controlled for efficient performance even under heavy loads
  • Digital display with speed and time setting
  • Precise RPM setting from 500 to 2200 min-¹ with lowest count of 10 min-¹
  • Enhanced airflow design to keep the temperature under control
  • Lid lock safety feature – lid opens automatically when run is completed
  • Timer setting from 1 to 999 minutes and infinite mode
  • One touch RPM / RCF conversion and short spin operation option on keypad
  • Compact design with low loading height for easy operation
Ordering information: Supplied with standard accessories.