VWR®, Autoclavable Rotors for Benchtop Centrifuge, Versatile Lab Genius
Supplier: VWR Collection
Certificates
About this item
Specifications
- Catalog No:
- VWRA521-2859
- VWRA521-2860
- VWRA521-2861
- VWRA521-2872
- VWRA521-2863
- VWRA521-2864
- VWRA521-2865
- VWRA521-2867
- VWRA521-2857
- VWRA521-2868
- VWRA521-2858
- Max. capacity:
- 6×50 ml
- 16×15 ml
- 12×15 ml
- 32×6 ml
- 2×microplate
- 4×100 ml
- 4×50 ml
- 24×15 ml
- 4×100 ml
- 16×15 ml
- 4×50 ml
- Max. RCF:
- 3485 xg
- 3164 xg
- 2355 xg
- 2599 xg
- 2644 xg
- 2854 xg
- 3164 xg
- 3144 xg
- 3485 xg