Order Entry
India
ContactUsLinkComponent
D-(-)-Sorbitol 91.0-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
D-(-)-Sorbitol 91.0-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Catalog # 3449-K1
Supplier:  Avantor
CAS Number:  
undefined
D-(-)-Sorbitol 91.0-100.5% (by anhydrous basis) NF, Multi-Compendial, J.T.Baker®
Catalog # 3449-K1
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
 :182.17 g/mol
CAS Number:50-70-4
 :98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708