Bloodborne pathogen and chemical splash protection lab coats KLEENGUARD* A60
Catalog # KIMB45512-10
Supplier: KIMBERLY CLARK
Specifications
- Clothing Size:M
- Garment application:
- Gender:
- Colour:Blue
- Description:Protection lab coats
- Environmentally Preferable:Waste Reducing
- Pk:25
Specifications
About this item
These three layer fabric bloodborne pathogens and chemical splash protection lab coat protects against exposure to blood and chemicals. These are made of microporous film to escape heat and sweat.
- Front snap closure
- One chest pocket in front
- One hip pocket in front
The typical applications are medical applications, animal research, food processing, liquid handling etc...
Certifications: ASTM F1670/1671‡ testing for penetration of blood, body fluids and bloodborne pathogens