Laboratory coats, breathable particle protection, KLEENGUARD® A20
Catalog # KIMB10039
Supplier: KIMBERLY CLARK
Specifications
- Clothing Size:XL
- Closure type:Press studs / snaps
- Pockets:With pockets
- Colour:White
- Description:Laboratory coat, KleenGuard™ A20
- Pk:25
Specifications
About this item
These KleenGuard™ personal protective equipment (PPE) lab coats are designed to help protect against hazardous and non-hazardous dust, dirt, grime and are made with breathable Microforce barrier fabric.
- Features serged seams, four front snap closures and open wrists
- Knee-length lab coats have a left chest pocket and right hip pocket
These protective garments offer superior particle holdout coupled with unmatched comfort attributes. The A20 fabric is comprised of five layers of stacked barrier protection (SMMMS) against hazardous particles. They are a popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments.
Certifications: NFPA 99 Compliant Antistatic Material.