Order Entry
India
ContactUsLinkComponent
Critical task wipers, KIMTECH PREP® SCOTTPURE™
  KIMB06121
undefined
Critical task wipers, KIMTECH PREP® SCOTTPURE™
  KIMB06121
  • Packed:
    100 sheets/pack, 4 packs/case
  • Colour:
    White
  • Description:
    SCOTTPURE™ Crtical task wipers, quarter-fold
  • L×W:
    305×381 mm
  • Pk:
    4
KIMTECH PREP® SCOTTPURE™ wipers are designed for wiping tasks where precision and reliability are essential.
  • Absorbent, low-lint polyester/rayon material has low extractables
  • Cloth-like wipers are acid-, base- and solvent-resistant
  • Box format helps protect the wipers before use
These critical task wipers are engineered for excellent absorbency and low-lint wiping. No binders or surfactants. Applications include aerospace, clean manufacturing areas, DVD manufacturing and printing.