Order Entry
India
ContactUsLinkComponent
di-Potassium hydrogen phosphate USP, Multi-Compendial, J.T.Baker®
  3250-X2
 :  Avantor
 :  
di-Potassium hydrogen phosphate USP, Multi-Compendial, J.T.Baker®
  3250-X2
 :  Avantor
 :  
  • Pack type:
    Bag/kit
  • Pk:
    68,038 kg
  • Seq:
    4496
  • Kit Components Meet Requirements:
    Passes Test
 :
 :174.18 g/mol
 :13,07743
 :340 °C
 :2,3 g/cm³ (20 °C)
 :Management of Change (MOC) category = HR
 :Ambient
 :231-834-5
 :7758-11-4
 :K₂HPO₄
 :MFCD00011383