



n-Pentane ≥98% (by GC), UltimAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACRV557-10
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
- Pk Info:1×4 L
Specifications
Specification Test Results
- Assay (as n-pentane) (by GC):≥98.0 %
- Assay (total C₅ isomers):≥99.0 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.00010 %
- Water (H₂O)(by KF, coulometric):≤0.01 %
- Optical Absorbance (1-cm path vs water) - 190 nm:≤1.0
- Optical Absorbance (1-cm path vs water) - 210 nm:≤0.25
- Optical Absorbance (1-cm path vs water) - 220 nm:≤0.05
- Optical Absorbance (1-cm path vs water) - 250 nm:≤0.01
- Optical Absorbance (1-cm path vs water) - 270 - 400 nm:≤0.005
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- ECD Sensitive Impurities (as Heptachl or Epoxide) Single Peak (ng/L):≤10 ppb
- FID Sensitive Impurities (as 2-Octano l) Single Peak (µg/L):≤5 ppb
- ACS - Neat Solvent Front Characterization:Passes Test
Specification Test Results
About this item
:
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0,63 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
:Management of Change (MOC) category = NI
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498