Nitric acid 69 - 70% (Standard of SFE), Macron Fine Chemicals™
Catalog # MACRV471-R
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:1 lb
- Seq:7032
- Pk Info:1×0,45 L
Specifications
Specification Test Results
- Assay (HNO₃):69.0 - 70.0 %
- Appearance:Passes Test
- Arsenic and Antimony (as As):≤0.005 ppm
- Chloride (Cl):≤0.08 ppm
- Heavy Metals (as Pb):≤0.10 ppm
- Phosphate (PO₄):≤0.20 ppm
- Residue after Ignition:≤2 ppm
- Sulfate (SO₄):≤0.50 ppm
- Color (APHA):≤10
- Trace Impurities - Aluminum (Al):≤0.100 ppm
- Trace Impurities - Barium (Ba):≤0.500 ppm
- Trace Impurities - Beryllium (Be):≤0.500 ppm
- Trace Impurities - Bismuth (Bi):≤0.500 ppm
- Trace Impurities - Boron (B):≤0.050 ppm
- Trace Impurities - Cadmium (Cd):≤0.500 ppm
- Trace Impurities - Calcium (Ca):≤0.200 ppm
- Trace Impurities - Chromium (Cr):≤0.100 ppm
- Trace Impurities - Cobalt (Co):≤0.500 ppm
- Trace Impurities - Copper (Cu):≤0.050 ppm
- Trace Impurities - Gallium (Ga):≤0.100 ppm
- Trace Impurities - Germanium (Ge):≤0.100 ppm
- Trace Impurities - Gold (Au):≤0.100 ppm
- Trace Impurities - Iron (Fe):≤0.100 ppm
- Trace Impurities - Lead (Pb):≤0.100 ppm
- Trace Impurities - Lithium (Li):≤1.000 ppm
- Trace Impurities - Magnesium (Mg):≤0.300 ppm
- Trace Impurities - Manganese (Mn):≤0.200 ppm
- Trace Impurities - Molybdenum (Mo):≤0.300 ppm
- Trace Impurities - Nickel (Ni):≤0.050 ppm
- Trace Impurities - Niobium (Nb):≤0.500 ppm
- Trace Impurities - Potassium (K):≤0.300 ppm
- Trace Impurities - Silicon (Si):≤0.500 ppm
- Trace Impurities - Silver (Ag):≤0.100 ppm
- Trace Impurities - Sodium (Na):≤0.300 ppm
- Trace Impurities - Strontium (Sr):≤0.500 ppm
- Trace Impurities - Tantalum (Ta):≤0.500 ppm
- Trace Impurities - Thallium (Tl):≤0.500 ppm
- Trace Impurities - Tin (Sn):≤0.300 ppm
- Trace Impurities - Titanium (Ti):≤0.300 ppm
- Trace Impurities - Vanadium (V):≤0.300 ppm
- Trace Impurities - Zinc (Zn):≤0.300 ppm
- Trace Impurities - Zirconium (Zr):≤0.300 ppm
Specification Test Results
About this item
:
:63.01 g/mol
Merck Index:13,06608
Melting Pt:–42 °C
ADR:8,II
:Management of Change (MOC) category = R
UN:2031
:GHS03
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349