Order Entry
India
ContactUsLinkComponent
D-(+)-Trehalose dihydrate 97.0-102.0% (by HPLC), Multi-Compendial, J.T.Baker®
D-(+)-Trehalose dihydrate 97.0-102.0% (by HPLC), Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
D-(+)-Trehalose dihydrate 97.0-102.0% (by HPLC), Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Endotoxin Concentration, EU/g:
    ≤0.6
  • Arsenic (As):
    ≤0.25 ppm
  • Cadmium (Cd):
    ≤0.03 ppm
  • Mercury (Hg):
    ≤0.05 ppm
  • Lead (Pb):
    ≤0.08 ppm
  • Cobalt (Co):
    ≤0.08 ppm
  • Nickel (Ni):
    ≤0.33 ppm
  • Vanadium (V):
    ≤0.17 ppm
  • Silver (Ag):
    ≤0.17 ppm
  • Iridium (Ir):
    ≤0.17 ppm
  • Gold (Au):
    ≤1.67 ppm
  • Osmium (Os):
    ≤0.17 ppm
  • Palladium (Pd):
    ≤0.17 ppm
  • Platinum (Pt):
    ≤0.17 ppm
  • Rhodium (Rh):
    ≤0.17 ppm
  • Ruthenium (Ru):
    ≤0.17 ppm
  • Selenium (Se):
    ≤1.33 ppm
  • Thallium (Tl):
    ≤0.13 ppm
  • Barium (Ba):
    ≤11.67 ppm
  • Chromium (Cr):
    ≤18.33 ppm
  • Copper (Cu):
    ≤5.00 ppm
  • Lithium (Li):
    ≤4.17 ppm
  • Molybdenum (Mo):
    ≤25.00 ppm
  • Antimony (Sb):
    ≤1.50 ppm
  • Tin (Sn):
    ≤10.00 ppm
  • Aluminum (Al):
    ≤10.00 ppm
  • Residual Methanol:
    ≤500 ppm
  • Residual Ethanol:
    ≤833 ppm
  • Residual Isopropanol:
    ≤833 ppm
  • NF - Assay (HPLC):
    97.0 - 102.0 %
  • NF - Chloride (Cl):
    ≤0.0125 %
  • NF - Color and Clarity of Solution: Absorbance @ 720 nm:
    ≤0.050
  • NF - Color and Clarity of Solution, Absorption difference:
    ≤0.100
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Identification C:
    Passes Test
  • NF - Microbial Limits: TAMC (cfu/g):
    ≤100 cfu/g
  • NF - Microbial Limits: TYMC (cfu/g):
    ≤100 cfu/g
  • NF - Microbial Limits: E. coli:
    None Detected
  • NF - Microbial Limits: Salmonella:
    None Detected
  • NF - Nitrogen content:
    ≤0.005 %
  • NF - pH of 10% solution:
    4.5 - 6.5
  • NF - Related Substances - total peaks eluting before trehalose:
    ≤0.5 %
  • NF - Related Substances - total peaks eluting after trehalose:
    ≤0.5 %
  • NF - Residue on Ignition:
    ≤0.10 %
  • NF - Soluble starch:
    Passes Test
  • NF - Specific Optical Rotation, anhydrous [Å]²º^D (+):
    197 - 201 Degree
  • NF - Sulfate (SO₄):
    ≤0.0200 %
  • NF - Water (by Karl Fischer titration):
    9.0 - 11.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Assay (LC):
    97.0 - 102.0 %
  • EP/BP - Chloride (Cl):
    ≤125 ppm
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Microbial Limits: TAMC (cfu/g):
    ≤100 cfu/g
  • EP/BP - Microbial Limits: TYMC (cfu/g):
    ≤100 cfu/g
  • EP/BP - Microbial Limits: E. coli:
    None Detected
  • EP/BP - Microbial Limits: Salmonella:
    None Detected
  • EP/BP - pH of 10% solution:
    4.5 - 6.5
  • EP/BP - Related Substances - Impurity A:
    ≤0.5 %
  • EP/BP - Related Substances - Impurity B:
    ≤0.5 %
  • EP/BP - Related Substances - Unspecified Impurities, each:
    ≤0.05 %
  • EP/BP - Related Substances - Total Impurities:
    ≤1.0 %
  • EP/BP - Soluble starch:
    Passes Test
  • EP/BP - Specific Optical Rotation, anhydrous [Å]²º^D (+):
    197 - 201 Degree
  • EP/BP - Sulfate (SO₄):
    ≤200 ppm
  • EP/BP - Sulfated ash:
    ≤0.1 %
  • EP/BP - Water (by Karl Fischer:
    9.0 - 11.0 %
  • JP - Assay (LC):
    98.0 - 101.0 %
  • JP - Chloride (Cl):
    ≤0.018 %
  • JP - Dextrin, soluble starch and sulfite:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤5 ppm
  • JP - Identification 1:
    Passes Test
  • JP - Identification 2:
    Passes Test
  • JP - Identification 3:
    Passes Test
  • JP - Nitrogen:
    ≤0.005 %
  • JP - pH of 10% solution:
    4.5 - 6.5
  • JP - Related Substances - total peaks eluting before trehalose:
    ≤0.5 %
  • JP - Related Substances - total peaks eluting after trehalose:
    ≤0.5 %
  • JP - Residue on Ignition:
    ≤0.1 %
  • JP - Specific Optical Rotation, anhydrous [Å]²º^D (+):
    197 - 201 Degree
  • JP - Sulfate (SO₄):
    ≤0.024 %
  • JP - Water (by Karl Fischer titration):
    9.0 - 11.0 %
  • Appearance (white to off white powder):
    Passes Test
  • Conductivity, uS cm-1:
    ≤15
  • Reducing Sugars:
    Passes Test
  • Glucose Area%:
    ≤0.5 %
  • Microbial Testing - Staphylococcus aureus (USP):
    None Detected
  • Microbial Testing - Pseudomonas aeruginosa (USP):
    None Detected
  • Specific Optical Rotation, dihydrate [Å]²º^D (+):
    177.5 - 180.5 Degree

Specification Test Results

Frequently Bought Together