


Methanol absolute ≥99.9% (by GC, corrected for water content) for HPLC, for liquid chromatography, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
ADR:3,II
:Management of Change (MOC) category = NI
UN:1230
:GHS02
:GHS06
:GHS08
Specification Test Results
- ACS - Assay (CH₃OH) (by GC, corrected for water):≥99.9 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 243 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm:≤0.15
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.80
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 205 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤205
- Liquid Chromatography Suitability - Absorbance:Passes Test
- Liquid Chromatography Suitability - HPLC Gradient Elution:Passes Test
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Appearance:Passes Test
- Color (APHA):≤10
- Water (by KF, coulometric):≤0.007 %
- Residue after Evaporation:≤1.0 ppm
- Solubility in H₂O:Passes Test
- Carbonyl Compounds - Acetone:≤0.001 %
- Carbonyl Compounds - Formaldehyde:≤0.001 %
- Carbonyl Compounds - Acetaldehyde:≤0.001 %
- Titrable Acid (µeq/g):≤0.3
- Titrable Base (µeq/g):≤0.10
- Substances Darkened by H₂SO₄:Passes Test
- Substances Reducing Permanganate:Passes Test
Specification Test Results
Frequently Bought Together