
Sodium chloride 99.0-100.5% (dry basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7647-14-5
:
Formula:NaCl
MDL Number:MFCD00003477
:58.44 g/mol
:Management of Change (MOC) category = R
:GHS07
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Acidity or Alkalinity:Passes Test
- USP - Appearance of Solution:Passes Test
- USP - Aluminum (Al):≤0.2 ppm
- USP - Arsenic (As):≤1 ppm
- USP - Barium (Ba):Passes Test
- USP - Bromide (Br):≤100 ppm
- USP - Ferrocyanide:Passes Test
- USP - Iodides (I):Passes Test
- USP - Iron (Fe):≤2 ppm
- USP - Loss on Drying at 105°C:≤0.5 %
- USP - Magnesium and Alkaline Earth Metals (as Ca):≤100 ppm
- USP - Nitrite (NO₂) (Absorbance Value):≤0.01
- USP - Phosphate (PO₄):≤25 ppm
- USP - Potassium (K):≤500 ppm
- USP - Sulfate (SO₄):≤200 ppm
- USP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Aluminum (Al):≤0.2 ppm
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Bromide (Br):≤100 ppm
- EP/BP - Ferrocyanide:Passes Test
- EP/BP - Iodide (I):Passes Test
- EP/BP - Nitrite (NO₂):≤0.01
- EP/BP - Phosphate (PO₄):≤25 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Arsenic (As):≤1 ppm
- EP/BP - Barium (Ba):Passes Test
- EP/BP - Iron (Fe):≤2 ppm
- EP/BP - Magnesium and Alkaline Earth Metals:≤100 ppm
- EP/BP - Loss on Drying at 105°C:≤0.5 %
- EP/BP - Potassium (K):≤500 ppm
- EP/BP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- EP/BP - Endotoxin Concentration, IU/g:≤2.5
- JP - Assay (NaCl) (dried basis):99.0 - 100.5 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Clarity and Color of Solution:Passes Test
- JP - Bromide (Br):Passes Test
- JP - Ferrocyanide:Passes Test
- JP - Iodide (I):Passes Test
- JP - Iron (Fe):Passes Test
- JP - Magnesium and Alkaline Earth Metals:Passes Test
- JP - Phosphate (PO₄):Passes Test
- JP - Sulfates (SO₄):Passes Test
- JP - Heavy Metals (as Pb):≤3 ppm
- JP - Barium (Ba):Passes Test
- JP - Arsenic (As):≤2 ppm
- JP - Loss on Drying:≤0.5 %
Specification Test Results
Related Information
Frequently Bought Together