Order Entry
India
ContactUsLinkComponent
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Irritant
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Irritant
Citric acid monohydrate 99.5-100.5% (by anhydrous basis), granular USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • USP - Clarity of Solution:
    Passes Test
  • USP - Color of Solution:
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Water (H₂O):
    7.5 - 9.0 %
  • USP - Readily Carbonizable Substances:
    Passes Test
  • USP - Residue on Ignition:
    ≤0.1 %
  • USP - Sulfate (SO₄):
    ≤0.015 %
  • USP - Oxalic Acid:
    ≤0.036 %
  • USP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • EP/BP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Identification E:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Readily Carbonizable Substances:
    Passes Test
  • EP/BP - Oxalic Acid:
    ≤360 ppm
  • EP/BP - Sulfate (SO₄):
    ≤150 ppm
  • EP/BP - Water (H₂O):
    7.5 - 9.0 %
  • EP/BP - Ash (sulfated):
    ≤0.1 %
  • EP/BP - Endotoxin Concentration, IU/mg:
    ≤0.5
  • JP - Assay (C₆H₈O₇) (anhydrous basis):
    99.5 - 100.5 %
  • JP - Identification:
    Passes Test
  • JP - Sulfate (SO₄):
    ≤150 ppm
  • JP - Oxalic Acid:
    ≤360 ppm
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals (as Pb):
    ≤10 ppm
  • JP - Water (H₂O):
    7.5 - 9.0 %
  • JP - Readily Carbonizable Substances:
    Passes Test
  • JP - Residue on Ignition:
    ≤0.10 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together