


Sulphuric acid 10.0 N in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MACRH379-05
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE bottle, narrow mouth
- Pk:1 L
- Seq:5037
- Pk Info:1×1 L
Specifications
Specification Test Results
- Normality:9.95 - 10.05
Specification Test Results
About this item
:
:98.08 g/mol
ADR:8,II
:Management of Change (MOC) category = N
EINECS:231-639-5
UN:2796
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589