

Phenol solution, StandARd®, Macron Fine Chemicals™
MACRH302-01
:
- Pack type:Glass bottle, amber, narrow mouth
- Pk:100 mL
- Seq:2440
- Pk Info:1×100 ml
- Concentration (0.8-1.2 mg/mL):Passes Test
:
:94.11 g/mol
:6.1,II
:Management of Change (MOC) category = N
:1671
:GHS07
:GHS08
:108-95-2
:C₆H₅OH
:MFCD00002143