Order Entry
India
ContactUsLinkComponent
Oxalic acid 10% (w/v) in aqueous solution, StandARd® (APHA), Macron Fine Chemicals™
Corrosive
Oxalic acid 10% (w/v) in aqueous solution, StandARd® (APHA), Macron Fine Chemicals™
Catalog # MACRH281-05
CAS Number:  
undefined
Corrosive
Oxalic acid 10% (w/v) in aqueous solution, StandARd® (APHA), Macron Fine Chemicals™
Catalog # MACRH281-05
CAS Number:  

Specifications

  • Pack type:
    PE bottle, narrow mouth
  • Pk:
    1 L
  • Seq:
    5037
  • Pk Info:
    1×1 L

Specifications

Specification Test Results

  • Assay (H₂C₂O₄·2H₂O)(g/100 mL):
    9.5 - 10.5

Specification Test Results

About this item

 :
 :90.04 g/mol
Density:1,02 g/cm³
ADR:8,III
 :Management of Change (MOC) category = N
UN:3265
 :GHS05
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573