Order Entry
India
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (dry basis) USP, Multi-Compendial, J.T.Baker®
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (dry basis) USP, Multi-Compendial, J.T.Baker®
  LP10-S1
 :  Avantor
 :  
undefined
Sodium dihydrogen phosphate monohydrate 98.0-103.0% (dry basis) USP, Multi-Compendial, J.T.Baker®
  LP10-S1
 :  Avantor
 :  

 

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645
  • Pk Info:
    1×500 g

 

 

  • Appearance (White crystals or granules):
    Passes Test
  • USP - Assay (anhydrous basis):
    98.0 - 103.0 %
  • USP - Aluminum, Calcium, and Related Elements:
    Passes Test
  • USP - Arsenic (As):
    ≤ 8 ppm
  • USP - Chloride (Cl):
    ≤ 0.014 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.2 %
  • USP - pH (1 in 20):
    4.1 - 4.5
  • USP - Sulfate (SO₄):
    ≤ 0.15 %
  • USP - Water (H₂O):
    10.0 - 15.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Identification C:
    Passes Test
  • EP - Assay (anhydrous basis):
    98.0 - 100.5 %
  • EP - Chloride (Cl):
    ≤ 200 ppm
  • EP - Sulfate (SO₄):
    ≤ 300 ppm
  • EP - Reducing Substances:
    Passes Test
  • Endotoxin Concentration (EU/g):
    ≤ 2.5
  • EP - Iron (Fe):
    ≤ 10 ppm
  • EP - Appearance of Solution:
    Passes Test
  • EP - pH:
    4.2 - 4.5
  • BP - Identification A:
    Passes Test
  • BP - Identification B:
    Passes Test
  • BP - Identification C:
    Passes Test
  • BP - Identification D:
    Passes Test
  • BP - Acidity:
    4.2 - 4.5
  • BP - Clarity and Color of Solution:
    Passes Test
  • BP - Arsenic (As):
    ≤ 2 ppm
  • BP - Iron (Fe):
    ≤ 10 ppm
  • BP - Chloride (Cl):
    ≤ 200 ppm
  • BP - Sulfate (SO₄):
    ≤ 300 ppm
  • BP - Reducing Substances:
    Passes Test
  • BP - Loss on Drying at 130°C:
    11.5 - 14.5 %
  • BP - Assay (dried basis):
    98.0 - 100.5 %
  • JPE - Assay (dried basis):
    ≥ 98.0 %
  • JPE - Identification:
    Passes Test
  • JPE - pH:
    4.1 - 4.7
  • JPE - Clarity and Color of Solution:
    Passes Test
  • JPE - Chloride (Cl):
    ≤ 0.005 %
  • JPE - Sulfate (SO4):
    ≤ 0.019 %
  • JPE - Heavy Metals:
    ≤ 10 ppm
  • JPE - Arsenic (As):
    ≤ 2 ppm

 

 

 :
 :137.99 g/mol
 :13,08734
 :100 °C
 :2,04 g/cm³
 :Management of Change (MOC) category = HR
 :Ambient
 :231-449-2
 :01-2119489796-13
 :10049-21-5
 :NaH₂PO₄·H₂O
 :MFCD00149208