Order Entry
India
ContactUsLinkComponent
Sodium hydroxide solution 0.5 N, Biotech Reagent, J.T.Baker®
Corrosive
Irritant
Sodium hydroxide solution 0.5 N, Biotech Reagent, J.T.Baker®
Catalog # BS26-02
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
Irritant
Sodium hydroxide solution 0.5 N, Biotech Reagent, J.T.Baker®
Catalog # BS26-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Pk Info:
    1×1 L

Specifications

Specification Test Results

  • Appearance (clear, colorless solution):
    Passes Test
  • Normality:
    0.480 - 0.520
  • Chloride (Cl):
    ≤5 ppm
  • ACS - Heavy Metals (as Pb):
    ≤1 ppm
  • Trace Impurities - Iron (Fe):
    ≤0.5 ppm
  • Endotoxin Concentration (EU/mL):
    ≤2.0

Specification Test Results

About this item

 :
 :40 g/mol
ADR:8,III
 :Management of Change (MOC) category = R
UN:1824
 :GHS05
 :GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548