

Sodium hydroxide 9.5 - 10.4 N in aqueous solution, Biotech Reagent, J.T.Baker®
Catalog # 5002-29
Supplier: Avantor
CAS Number:
Specifications
- Pk:800 L
- Seq:5693
- Pk Info:1×800 L
Specifications
Specification Test Results
- Identification:Passes Test
- Appearance (Clear, colorless liquid):Passes Test
- Normality:9.5 - 10.4
Specification Test Results
About this item
:
:40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1,53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
:Management of Change (MOC) category = R
:GHS05
:GHS07
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548