Order Entry
India
ContactUsLinkComponent
Magnesium chloride hexahydrate 99.0-105.0% FCC, Macron Fine Chemicals™
Magnesium chloride hexahydrate 99.0-105.0% FCC, Macron Fine Chemicals™
Catalog # MACR5933-24
CAS Number:  
undefined
Magnesium chloride hexahydrate 99.0-105.0% FCC, Macron Fine Chemicals™
Catalog # MACR5933-24
CAS Number:  

Specifications

  • Pk:
    100 lb
  • Seq:
    7167
  • Pk Info:
    1×45,36 kg

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • FCC - Assay (MgCl₂.6H₂O):
    99.0 - 105.0 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Ammonium (NH₄):
    ≤0.005 %
  • FCC - Lead (Pb):
    ≤4 mg/kg
  • FCC - Sulfate (SO₄):
    ≤0.03 %
  • Solubility:
    Passes Test

Specification Test Results

About this item

 :
 :203.3 g/mol
Melting Pt:117 °C
 :Management of Change (MOC) category = RL
CAS Number:7791-18-6
Formula:MgCl₂·6H₂O
MDL Number:MFCD00149781