Sulphuric acid 96%, Finyte®, J.T.Baker®
Catalog # 5815-54
Supplier: Avantor
CAS Number:
Specifications
- Pk:750 lb
- Seq:7368
- Environmentally Preferable:
- Pk Info:1×340,2 kg
Specifications
Specification Test Results
- Assay (H₂SO₄):95.0 - 97.0 %
- Color (APHA):≤10
- Chloride (Cl):≤0.1 ppm
- Nitrate (NO₃):≤0.1 ppm
- Phosphate (PO₄):≤0.20 ppm
- Trace Impurities - Aluminum (Al):≤4.5 ppb
- Arsenic and Antimony (as As):≤2.0 ppb
- Trace Impurities - Barium (Ba):≤2.0 ppb
- Trace Impurities - Beryllium (Be):≤1.0 ppb
- Trace Impurities - Bismuth (Bi):≤10.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤5.0 ppb
- Trace Impurities - Calcium (Ca):≤9.0 ppb
- Trace Impurities - Chromium (Cr):≤1.7 ppb
- Trace Impurities - Cobalt (Co):≤1.8 ppb
- Trace Impurities - Copper (Cu):≤2.5 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤5.0 ppb
- Trace Impurities - Iron (Fe):≤8.2 ppb
- Trace Impurities - Lead (Pb):≤2.0 ppb
- Trace Impurities - Lithium (Li):≤1.1 ppb
- Trace Impurities - Magnesium (Mg):≤1.8 ppb
- Trace Impurities - Manganese (Mn):≤1.7 ppb
- Trace Impurities - Molybdenum (Mo):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤2.0 ppb
- Trace Impurities - Niobium (Nb):≤5.0 ppb
- Trace Impurities - Potassium (K):≤3.7 ppb
- Trace Impurities - Silicon (Si):≤10.0 ppb
- Trace Impurities - Silver (Ag):≤5.0 ppb
- Trace Impurities - Sodium (Na):≤6.9 ppb
- Trace Impurities - Strontium (Sr):≤5.0 ppb
- Trace Impurities - Tantalum (Ta):≤5.0 ppb
- Trace Impurities - Thallium (Tl):≤5.0 ppb
- Trace Impurities - Tin (Sn):≤5.0 ppb
- Trace Impurities - Titanium (Ti):≤1.7 ppb
- Trace Impurities - Vanadium (V):≤5.0 ppb
- Trace Impurities - Zinc (Zn):≤3.4 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count at point of fill - 0.2 µm and greater:≤4000 par/ml
- Particle Count at point of fill - 0.5 µm and greater:≤300 par/ml
- Particle Count at point of fill - 1.0 µm and greater:≤60 par/ml
- Particle Count at point of fill - 0.1 µm and greater (Rion KS40AF):≤7635 par/ml
- Particle Count, finished unit - 0.1 µm and greater(Rion KS40AF) (FUT):≤6900 par/ml
Specification Test Results
About this item
:
:98.08 g/mol
ADR:8,II
:Management of Change (MOC) category = R
UN:1830
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589