Sodium thiosulphate 0.20 N (N/5), StandARd® volumetric solution, Macron Fine Chemicals™
Catalog # MACR5531-60
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE bottle, narrow mouth
- Pk:1 L
- Seq:5037
- Pk Info:1×1 L
Specifications
Specification Test Results
- Normality:0.199 - 0.201
Specification Test Results
About this item
:
:158.11 g/mol
Density:1,06 g/cm³
:Management of Change (MOC) category = N
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499