


4-Methyl-2-pentanone ≥99% (by GC, corrected for water content), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACR5923-08
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Pk Info:1×4 L
Specifications
Specification Test Results
- ACS - Color:≤15
- ACS - Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water):≥99 %
- ACS - Residue after Evaporation:≤5 ppm
- ACS - Titrable Acid (meq/g):≤0.002
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.08 %
- Optical Absorbance (1-cm path vs water) - 335 nm:≤1.00
- Optical Absorbance (1-cm path vs water) - 340 nm:≤0.50
- Optical Absorbance (1-cm path vs water) - 350 nm:≤0.25
- Optical Absorbance (1-cm path vs water) - 360 nm:≤0.15
- Optical Absorbance (1-cm path vs water) - 400 nm:≤0.01
Specification Test Results
About this item
:
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117,4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0,801 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:Management of Change (MOC) category = N
:GHS02
:GHS07
:GHS08
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938