


Methanol ≥99.5% (by GC), Macron Fine Chemicals™
MACR8820-28
:
- Pk:350 lb
- Seq:7251
- Pk Info:1×4 L
- NF - Assay (CH₃OH) (by GC):≥99.5 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Acidity:Passes Test
- NF - Alkalinity (as NH₃):≤3 ppm
- NF - Nonvolatile Residue:≤0.001 %
- NF - Readily Carbonizable Substances:Passes Test
- NF - Readily Oxidizable Substances:Passes Test
- NF - Acetone, Aldehydes (as (CH₃)₂CO):≤0.003 %
- NF - Water (H₂O):≤0.1 %
:
:32.04 g/mol
:3,II
:Management of Change (MOC) category = N
:1230
:GHS02
:GHS06
:GHS08
:67-56-1
:CH₃OH
:MFCD00004595