Order Entry
India
ContactUsLinkComponent
Bismuth(III) nitrate Liquor 51 Intermediate, Macron Fine Chemicals™
Oxidizing
Corrosive
Bismuth(III) nitrate Liquor 51 Intermediate, Macron Fine Chemicals™
Catalog # MACR9780-88
CAS Number:  
undefined
Oxidizing
Corrosive
Bismuth(III) nitrate Liquor 51 Intermediate, Macron Fine Chemicals™
Catalog # MACR9780-88
CAS Number:  

Specifications

  • Pk:
    220 gal US
  • Seq:
    8275
  • Pk Info:
    1×832,79 L

Specifications

Specification Test Results

  • Assay (as Bi):
    20.0 - 23.0 %
  • Free Nitric Acid Assay (g/L):
    70.0 - 90.0 g/l
  • Specific Gravity:
    1.52 - 1.58

Specification Test Results

About this item

 :
 :395
ADR:8, 5.1,II
 :Management of Change (MOC) category = N
UN:3093
 :GHS03
 :GHS05
CAS Number:10361-44-1
Formula:Bi(NO₃)₃·xH₂O
MDL Number:MFCD00003460