


Aniline ≥99.0% (by GC, corrected for water content), AR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:62-53-3
:
Formula:C₆H₅NH₂
MDL Number:MFCD00007629
:93.13 g/mol
Merck Index:13,00661
Boiling Pt:184 °C (1013 hPa)
Melting Pt:–6,2 °C
Density:1,019 g/cm³ (20 °C)
EINECS:200-539-3
UN:1547
ADR:6.1,II
:Management of Change (MOC) category = N
Flash Pt:76 °C
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Assay (C₆H₅NH₂) (by GC, corrected for water):≥99.0 %
- Color (APHA) (at time of packaging):≤250
- Residue after Ignition:≤0.005 %
- Chlorobenzene (C₆H₅Cl):≤0.01 %
- Hydrocarbons:Passes Test
- Nitrobenzene (C₆H₅NO₂):Passes Test
Specification Test Results
Frequently Bought Together