


Acetone ≥99.5% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:67-64-1
:
Formula:CH₃COCH₃
MDL Number:MFCD00008765
:58.08 g/mol
Merck Index:13,00067
Boiling Pt:56,2 °C (1013 hPa)
Melting Pt:–95,4 °C
Density:0,792 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
:Management of Change (MOC) category = NI
Flash Pt:<–20 °C (closed cup)
REACH:01-2119471330-49
:GHS02
:GHS07
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- ACS - Assay ((CH₃)₂CO) (by GC, corrected for water):≥99.5 %
- ACS - Aldehydes (as HCHO):≤0.002 %
- ACS - Color (APHA):≤10
- ACS - ALDOL Condensation Products:≤800 ppm
- ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC):≤0.05 %
- ACS - Methanol (CH₃OH)(by GC):≤0.05 %
- ACS - Residue after Evaporation:≤0.0001 %
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Titrable Acid (µeq/g):≤0.3
- ACS - Titrable Base (µeq/g):≤0.6
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:≤0.06
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:≤0.01
- Water (H₂O):≤0.5 %
- Fluorescence Trace Impurities, in ppb, measured as Quinine Base::≤1.0 ppb
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
Specification Test Results
Frequently Bought Together