Order Entry
India
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.0-101.0% USP, Multi-Compendial, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • USP - Assay (C₄H₁₁NO₃):
    99.0 - 101.0 %
  • USP - Identification A:
    Passes Test
  • USP - Identification C:
    Passes Test
  • USP - pH (1 in 20):
    10.0 - 11.5
  • USP - Loss on Drying:
    ≤ 1.0 %
  • USP - Residue on Ignition:
    ≤ 0.1 %
  • USP - Melting Range(Lower):
    168 - 172 °C
  • USP - Melting Range(Upper):
    168 - 172 °C
  • EP/BP - Assay (C₄H₁₁NO₃) (dried basis):
    99.0 - 100.5 %
  • EP/BP - Identification B (melting point):
    168 - 174 °C
  • EP/BP - Identification C:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - pH:
    10.0 - 11.5
  • EP/BP - Related Substances:
    ≤ 1.0 %
  • EP/BP - Chloride (Cl):
    ≤ 100 ppm
  • EP/BP - Iron (Fe):
    ≤ 10 ppm
  • EP/BP - Loss on Drying at 105°C:
    ≤ 0.5 %
  • EP/BP - Ash (sulfated):
    ≤ 0.1 %
  • JPC 1997 - Assay (C₄H₁₁NO₃) (dried basis):
    ≥ 99.0 %
  • JPC 1997 - Identification A:
    Passes Test
  • JPC 1997 - Identification B:
    Passes Test
  • JPC 1997 - pH:
    10.3 - 10.7
  • JPC 1997 - Melting Point:
    168 - 172 °C
  • JP - Clarity and Color of Solution:
    Passes Test
  • JPC 1997 - Heavy Metals (as Pb):
    ≤ 8 ppm
  • JPC 1997 - Arsenic (As):
    ≤ 1.6 ppm
  • JPC 1997 - Loss on Drying:
    ≤ 0.5 %
  • JPC 1997 - Residue on Ignition:
    ≤ 0.10 %

Specification Test Results

Related Information

Frequently Bought Together