Sodium acetate, anhydrous 99.0-101.0% (dry basis) FCC, USP, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:127-09-3
:
Formula:H₃CCOONa
MDL Number:MFCD00012459
:82.03 g/mol
Merck Index:12,08711
Boiling Pt:881,4 °C (1013 hPa)
Melting Pt:324 °C
Density:1,528 g/cm³ (20 °C)
:Management of Change (MOC) category = R
Flash Pt:>250 °C (closed cup)
Storage Temperature:Ambient
EINECS:204-823-8
REACH:01-2119485123-42
Specification Test Results
- USP - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- USP - Calcium and Magnesium:Passes Test
- USP - Chloride (Cl):≤0.035 %
- USP - Heavy Metals (as Pb):≤10 ppm
- USP - Identification:Passes Test
- USP - Insoluble Matter:≤0.05 %
- USP - Loss on Drying at 120°C:≤1.0 %
- USP - pH of 3% Solution at 25°C:7.5 - 9.2
- USP - Potassium Compounds:Passes Test
- USP - Sulfate (SO₄):≤0.005 %
- FCC - Assay (CH₃COONa) (dried basis):99.0 - 101.0 %
- FCC - Alkalinity:≤0.2 %
- FCC - Identification:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Loss on Drying at 120°C:≤1.0 %
- FCC - Potassium Compounds:Passes Test
Specification Test Results
Frequently Bought Together