Order Entry
India
ContactUsLinkComponent
Ammonium sulphate ≥99.0%, granular ACS, FCC
Health
Ammonium sulphate ≥99.0%, granular ACS, FCC
Catalog # 3514-65
Supplier:  Avantor
CAS Number:  
undefined
Health
Ammonium sulphate ≥99.0%, granular ACS, FCC
Catalog # 3514-65
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    HDPE drum, Flowmor™ package
  • Pk:
    100 kg
  • Seq:
    4553
  • Pk Info:
    1×100 kg

Specifications

Specification Test Results

  • ACS - Assay ((NH₄)₂SO₄) (A.C.S.):
    ≥99.0 %
  • ACS - Chloride (Cl):
    ≤3 ppm
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Iron (Fe):
    ≤5 ppm
  • ACS - Nitrate (NO₃):
    ≤0.001 %
  • ACS - pH of 5% Solution at 25°C:
    5.0 - 6.0
  • ACS - Phosphate (PO₄):
    ≤3 ppm
  • ACS - Residue on Ignition:
    ≤0.005 %
  • Heavy Metals (as Pb):
    ≤3 ppm
  • Arsenic (As):
    ≤0.200 ppm
  • Solution Test:
    Passes Test
  • FCC - Assay ((NH₄)₂SO₄)(F.C.C.):
    99.0 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Lead (Pb):
    ≤3 mg/kg
  • FCC - Residue on Ignition (FCC):
    ≤0.25 %
  • FCC - Selenium (Se):
    ≤0.003 %
  • Loss on Drying at 105°C:
    ≤0.1 %

Specification Test Results

About this item

 :
 :132.14 g/mol
 :Management of Change (MOC) category = RL
Flash Pt:1,77 g/cm³
 :GHS08
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391