Sulphuric acid 96%, Microelectronic Grade, Macron Fine Chemicals™
MACR2877-R
:
- Pk:1 lb
- Seq:7032
- Environmentally Preferable:
- Pk Info:1×1 kg
- Assay (H₂SO₄) (by acidimetry):95.0 - 97.0 %
- Color (APHA):≤8
- Residue after Ignition:≤2 ppm
- Chloride (Cl):≤0.1 ppm
- Substances Reducing Permanganate (as SO₂):Passes Test
- Arsenic and Antimony (as As):≤0.005 ppm
- Phosphate (PO₄):≤0.5 ppm
- Nitrate (NO₃):≤0.2 ppm
- Trace Impurities - Aluminum (Al):≤0.200 ppm
- Trace Impurities - Barium (Ba):≤0.300 ppm
- Trace Impurities - Boron (B):≤0.010 ppm
- Trace Impurities - Cadmium (Cd):≤0.500 ppm
- Trace Impurities - Calcium (Ca):≤0.300 ppm
- Trace Impurities - Chromium (Cr):≤0.200 ppm
- Trace Impurities - Cobalt (Co):≤0.500 ppm
- Trace Impurities - Copper (Cu):≤0.10 ppm
- Trace Impurities - Gallium (Ga):≤0.500 ppm
- Trace Impurities - Germanium (Ge):≤0.500 ppm
- Trace Impurities - Gold (Au):≤0.300 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Trace Impurities - Lead (Pb):≤0.300 ppm
- Trace Impurities - Lithium (Li):≤0.500 ppm
- Trace Impurities - Magnesium (Mg):≤0.300 ppm
- Trace Impurities - Manganese (Mn):≤0.200 ppm
- Trace Impurities - Nickel (Ni):≤0.1 ppm
- Trace Impurities - Potassium (K):≤0.300 ppm
- Trace Impurities - Silicon (Si):≤0.500 ppm
- Trace Impurities - Silver (Ag):≤0.300 ppm
- Trace Impurities - Sodium (Na):≤0.300 ppm
- Trace Impurities - Strontium (Sr):≤0.300 ppm
- Trace Impurities - Tin (Sn):≤0.200 ppm
- Trace Impurities - Titanium (Ti):≤0.300 ppm
- Trace Impurities - Zinc (Zn):≤0.200 ppm
:
:98.08 g/mol
:231-639-5
:1830
:GHS05
:GHS07
:GHS08
:Ambient
:7664-93-9
:H₂SO₄
:MFCD00064589
:∼335 °C (1013 hPa)
:3 °C
:1.84 g/cm³ (20 °C)
:8 II
:14,08974
:Management of Change (MOC) category = R