Phosphomolybdic acid, crystals, AR® ACS, Macron Fine Chemicals™
Catalog # MACR2772-02
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE bottle, Mall Polystormor®
- Pk:125 g
- Seq:3445
- Pk Info:1×125 g
Specifications
Specification Test Results
- ACS - Appearance (bright yellow granular crystals turning to bright yellow-green):Passes Test
- ACS - Ammonium (NH₄):≤0.01 %
- ACS - Calcium (Ca):≤0.02 %
- ACS - Chloride (Cl):≤0.02 %
- ACS - Heavy Metals (as Pb):≤0.005 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Iron (Fe):≤0.005 %
- ACS - Sulfate (SO₄):≤0.025 %
Specification Test Results
About this item
:
ADR:8,III
:Management of Change (MOC) category = N
UN:1759
:GHS05
CAS Number:11104-88-4
Formula:H₃Mo₁₂O₄₀P·xH₂O