Hydrochloric acid 30%, Technical Grade for trace metal analysis (20° Baume), Macron Fine Chemicals™
Catalog # MACR2624-44
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, clear
- Pk:2,5 L
- Seq:5096
- Pk Info:1×2,5 L
Specifications
Specification Test Results
- Assay (HCl):≥34 %
- Identification:Passes Test
- Arsenic (As):≤5 ppm
- Free Chlorine (as Cl):≤1 ppm
- Nitrate (NO₃):≤0.005 %
- Other Heavy Metals (as Pb):≤1 ppm
- Total Iron (Fe):≤10 ppm
Specification Test Results
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1,18 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:1789
ADR:8,II
:Management of Change (MOC) category = N
:36.46 g/mol
EINECS:231-595-7
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324