Paraformaldehyde, OR, Macron Fine Chemicals™
Catalog # MACR2621-59
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
- Pk Info:1× 500 g
Specifications
Specification Test Results
- Identification:Passes Test
Specification Test Results
About this item
Synonyms: Polyoxymethylene
:
ADR:4.1,III
:Management of Change (MOC) category = N
UN:2213
CAS Number:30525-89-4
Formula:(CH₂O)n
MDL Number:MFCD00133991