Silver chloride, AR®, Macron Fine Chemicals™
Catalog # MACR2147-03
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:16 oz
- Seq:8515
- Pk Info:1× 453 g (16 oz)
Specifications
Specification Test Results
- Solubility:Passes Test
- Copper (Cu):≤0.001 %
- Heavy Metals (as Pb):≤0.005 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Nitrate (NO₃):≤0.001 %
Specification Test Results
About this item
:
:143.32 g/mol
Merck Index:13,08582
Boiling Pt:1554 °C (1013 hPa)
Melting Pt:455 °C
Density:5,56 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-033-3
UN:3077
ADR:9,III
:Management of Change (MOC) category = N
:GHS06
:GHS09
CAS Number:7783-90-6
Formula:AgCl
MDL Number:MFCD00003399