Glycerine (vegetable based) 98.0-101.0% (by anhydrous basis), Biotech Reagent low in aldehydes, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Density:1,26 g/cm³
:Management of Change (MOC) category = N
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B - Diethylene Glycol:≤0.10 %
- USP - Identification B - Ethylene Glycol:≤0.10 %
- USP - Identification C:Passes Test
- USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis):99.0 - 101.0 %
- USP - Specific Gravity at 25°/25°C:≥1.249
- USP - Color:Passes Test
- USP - Residue on Ignition:≤0.01 %
- USP - Chloride (Cl):≤0.001 %
- USP - Sulfate (SO₄):≤0.002 %
- USP - Chlorinated Compounds (as Cl):≤0.003 %
- USP - Fatty Acids and Esters:Passes Test
- USP - Heavy Metals (as Pb):≤5 ppm
- USP - Water (H₂O):≤2.0 %
- USP - Related Compounds: Individual Impurity:≤0.1 %
- USP - Related Compo unds: Total Impurities:≤1.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Refractive Index, n²ºD:1.470 - 1.475
- EP/BP - Aldehydes:≤10 ppm
- EP/BP - Ester:Passes Test
- EP/BP - Halogenated Compounds:≤35 ppm
- EP/BP - Sugars:Passes Test
- EP/BP - Chloride (Cl):≤10 ppm
- EP/BP - Heavy Metals (as Pb):≤5 ppm
- EP/BP - Water (H₂O):≤2.0 %
- EP/BP - Ash (sulfated):≤0.01 %
- EP/BP - Impurity A and Related Substances - Impurity A:≤0.1 %
- EP/BP - Impurity A and Related Substances - Other Impurities with R^T≤0.1 %
- EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol:≤0.5 %
- EP - Assay (anhydrous basis):98.0 - 101.0 %
- JP - Assay (anhydrous basis):98.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Specific Gravity at 20°/20°C:≥1.258
- JP - Refractive Index, n²ºD:≥1.470
- JP - Color:Passes Test
- JP - Residue on Ignition:≤0.01 %
- JP - Chloride (Cl):≤0.001 %
- JP - Sulfate (SO₄):≤0.002 %
- JP - Heavy Metals (as Pb):≤5 ppm
- JP - Fatty Acids and Esters(as butyric acid):Passes Test
- JP - Readily Carbonizable Substances:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Acrolein, Glucose, and Other Reducing Substances:Passes Test
- JP - Ammonium (NH₄):Passes Test
- JP - Calcium (Ca):Passes Test
- JP - Arsenic (As):≤2 ppm
- JP - Water (H₂O):≤2.0 %
- JP - Ethylene Glycol:≤0.1 %
- JP - Diethylene Glycol:≤0.1 %
- JP - Any Other Impurity:≤0.1 %
- JP - Total Impurities:≤1.0 %
- Endotoxin Concentration (EU/g):≤5
- Microbial Limit: Total aerobic count:≤1000 cfu/g
- Microbial Limit: Total molds and yeasts:≤100 cfu/g
- Microbial Limit: Escherichia coli:None Detected
Specification Test Results
Frequently Bought Together