Order Entry
India
ContactUsLinkComponent
Tributyl phosphate, OR, Macron Fine Chemicals™
Health
Irritant
Tributyl phosphate, OR, Macron Fine Chemicals™
Catalog # MACR1940-61
CAS Number:  
Health
Irritant
Tributyl phosphate, OR, Macron Fine Chemicals™
Catalog # MACR1940-61
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg
  • Seq:
    4024
  • Pk Info:
    1× 1 kg

Specifications

Specification Test Results

  • Color (APHA):
    ≤50
  • Infrared Spectrum:
    Passes Test

Specification Test Results

About this item

 :
 :266.32 g/mol
Merck Index:13,09692
Boiling Pt:289 °C (1013 hPa)
Melting Pt:–79 °C
Density:0,982 g/cm³ (20 °C)
 :Management of Change (MOC) category = N
Flash Pt:151 °C
Storage Temperature:Ambient
EINECS:204-800-2
 :GHS08
 :GHS07
CAS Number:126-73-8
Formula:(CH₃(CH₂)₃O)₃PO
MDL Number:MFCD00009436