
Ammonium sulphate kit 99.0-100.5% NF endotoxin tested, J.T.Baker®
Catalog # 0798-X1
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Bag/kit
- Pk:1 KIT
- Seq:0075
- Pk Info:1× 50 kg + 1× 500 g
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1,77 g/cm³
EINECS:231-984-1
REACH:01-2119455044-46
:GHS08
Storage Temperature:Ambient
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391