Order Entry
India
ContactUsLinkComponent
Ammonium sulphate kit 99.0-100.5% NF endotoxin tested, J.T.Baker®
Health
Ammonium sulphate kit 99.0-100.5% NF endotoxin tested, J.T.Baker®
Catalog # 0798-X2
Supplier:  Avantor
CAS Number:  
Health
Ammonium sulphate kit 99.0-100.5% NF endotoxin tested, J.T.Baker®
Catalog # 0798-X2
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Bag/kit
  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    1× 100 kg + 1× 500g

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
 :132.14 g/mol
Merck Index:13,00559
Melting Pt:336…339 °C
Density:1,77 g/cm³
EINECS:231-984-1
REACH:01-2119455044-46
 :GHS08
Storage Temperature:Ambient
CAS Number:7783-20-2
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391