Magnesium chloride hexahydrate ≥98%, crystals USP, Multi-Compendial, J.T.Baker®
: Avantor
:7791-18-6
:
:MgCl₂·6H₂O
:MFCD00149781
:203.3 g/mol
:13,05684
:1412 °C (1013 hPa)
:117 °C
:1,57 g/cm³ (25 °C)
:Management of Change (MOC) category = HR
:Ambient
:232-094-6
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH (1 in 20):4.5 - 7.0
- USP - Insoluble Matter:≤ 0.005 %
- USP - Sulfate (SO₄):≤ 0.005 %
- USP - Barium (Ba):Passes Test
- USP - Calcium (Ca):≤ 0.01 %
- USP - Potassium (K):Passes Test
- USP - Assay:98.0 - 101.0 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Assay:98.0 - 101.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Bromide (Br):≤ 500 ppm
- EP/BP - Sulfate (SO₄):≤ 100 ppm
- EP/BP - Calcium (Ca):≤ 0.1 %
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Water (H₂O):51.0 - 55.0 %
Frequently Bought Together