di-Potassium hydrogen phosphate, anhydrous 98.0-100.5% (dried basis), Multi-Compendial, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7758-11-4
:
Formula:K₂HPO₄
MDL Number:MFCD00011383
:174.18 g/mol
Merck Index:13,07743
Melting Pt:340 °C
Density:2,3 g/cm³ (20 °C)
:Management of Change (MOC) category = HR
Storage Temperature:Ambient
EINECS:231-834-5
Specification Test Results
- USP - Assay (K₂HPO₄) (dried basis):98.0 - 100.5 %
- USP - Arsenic (As):≤ 3 ppm
- USP - Carbonate (CO₃):Passes Test
- USP - Chloride (Cl):≤ 0.03 %
- USP - Endotoxin Concentration (2.5 EU/g max.):Passes Test
- USP - Fluoride (F):≤ 0.001 %
- USP - Identification A (USP):Passes Test
- USP - Identification B (USP):Passes Test
- USP - Insoluble Substances:≤ 0.2 %
- USP - Iron (Fe):≤ 0.003 %
- USP - Loss on Drying:≤ 1.0 %
- USP - Mono- or Tri- Basic Salt:Passes Test
- USP - pH (1 in 20):8.5 - 9.6
- USP - Sodium (Na):Passes Test
- USP - Sulfate (SO₄):≤ 0.1 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Assay (K₂HPO₄) (dried basis):98.0 - 101.0 %
- EP/BP - Chloride (Cl):≤ 200 ppm
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Loss on Drying:≤ 2.0 %
- EP/BP - Monopotassium phosphate:≤ 0.025
- EP/BP - Reducing Substances:Passes Test
- EP/BP - Sodium (Na):≤ 0.1 %
- EP/BP - Sulfate (SO₄):≤ 0.1 %
Specification Test Results
Frequently Bought Together