Order Entry
India
ContactUsLinkComponent
Potassium thiocyanate ≥99.00% AR ACS, Rankem®
Irritant
Potassium thiocyanate ≥99.00% AR ACS, Rankem®
Catalog # RANKP0612
Supplier:  RANKEM
CAS Number:  
Irritant
Potassium thiocyanate ≥99.00% AR ACS, Rankem®
Catalog # RANKP0612
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay (Argentometry):
    Min. 99.00%
  • pH (5%, Water; at 25°C):
    5.30 --> 8.50
  • Water Insoluble Matter:
    Max. 0.005%
  • Chloride (Cl):
    ≤0.005%
  • Sulphate (SO4):
    ≤0.005%
  • Sulphide (S):
    ≤0.001%
  • Iron (Fe):
    ≤ 0.0001%
  • Ammonium (NH4):
    ≤0.003%
  • Heavy Metal (As Pb):
    ≤0.0005%
  • Copper(Cu):
    Max. 0.0002 %
  • Iodine Consuming Substances:
    Passes test
  • Sodium (Na):
    Max. 0.005%

Specification Test Results

About this item

 :
Merck Index:12,07866
Boiling Pt:500 °C (1013 hPa)
Melting Pt:175 °C
Density:1,61 g/cm³ (20 °C)
 :97.18 g/mol
Storage Temperature:Ambient
EINECS:206-370-1
 :GHS07
CAS Number:333-20-0
Formula:KSCN
MDL Number:MFCD00011413